• USA Home
  • S9438 - Somatostatin Fragment 3-10

EMAIL THIS PAGE TO A FRIEND
S9438 Sigma

Somatostatin Fragment 3-10

≥96% (HPLC), solid

  • Empirical Formula (Hill Notation) C52H72N12O11S

  • Molecular Weight 1073.27

  •  MDL number MFCD03093454

  •  PubChem Substance ID 329824624

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Peptides and Proteins, Peptides for Cell Biology,
InChI Key   NXKPSJBXWYFMKT-LWTFXWKPSA-N
assay   ≥96% (HPLC)
form   solid
storage temp.   −20°C
Gene Information   human ... SST(6750)

Description

Amino Acid Sequence

Cys-Lys-Asn-Phe-Phe-Trp-Lys-Thr

Price and Availability


Amplified Detection

Making Headway

All labs need water
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3
Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?