• USA Home
  • S9563 - Somatostatin Fragment 7-14

EMAIL THIS PAGE TO A FRIEND
S9563 Sigma

Somatostatin Fragment 7-14

≥95% (HPLC), solid

  • Empirical Formula (Hill Notation) C49H66N10O12S

  • Molecular Weight 1019.17

  •  MDL number MFCD03093453

Purchase

Properties

Related Categories Cell Biology, Cell Signaling and Neuroscience, Neuropeptides, Peptides and Proteins, Peptides for Cell Biology,
assay   ≥95% (HPLC)
form   solid
storage temp.   −20°C
Gene Information   human ... SST(6750)

Description

Amino Acid Sequence

Phe-Trp-Lys-Thr-Phe-Thr-Ser-Cys

Price and Availability


Amplified Detection

Amicon Ultra Centrifugal Filters
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?