• USA Home
  • S9641 - Hamilton® GASTIGHT® 1700 series syringe
EMAIL THIS PAGE TO A FRIEND
S9641 Sigma
Hamilton® GASTIGHT® 1700 series syringe
Model 1705TLL with PTFE Luer lock, capacity 50 μL
Purchase
Properties
Related Categories GASTIGHT, Hamilton syringes, Labware, Syringes and Needles
capacity   50 μL
Description
General description
For gas and liquid chromatography, corrosive gases, and volatile liquids. Chemically inert PTFE-tipped plunger. Leaktight syringe with 200 psi pressure rating. Cemented and removable needle models have 2 in. L, point style 2 needles.
Needles are not included with the Luer tip and TLL syringes.
*Indicates plunger is PTFE-coated.
Legal Information
GASTIGHT is a registered trademark of Hamilton Co.
Hamilton is a registered trademark of Hamilton Co.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
replaced by
Product #
Description
Add to Cart
26202 Hamilton® GASTIGHT® syringe, 1700 series, PTFE luer lock, 1705TLL, volume 50 μL, PTFE Luer lock, needle size (not included)
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?