• USA Home
  • S9685 - Structure-Function Analysis of G Protein-Coupled Receptors
EMAIL THIS PAGE TO A FRIEND
S9685 Sigma
Structure-Function Analysis of G Protein-Coupled Receptors
  •  ISBN-10 0-471-25228-X
  •  ISBN-13 978-0-471-25228-3
Purchase
Properties
Related Categories Books and Software, Cell Signaling Books, Labware More...
publication info   J. Wess, ed., Wiley-Liss, 1999, 412 pp., hard cover
mfr. no.   Wiley
Description
General description
G Protein Coupled Receptors (GPCR) often play key roles in a large number of physiological and pathophysiological conditions. Theses receptors are the targets of a large number of therapeutic drugs. Detailed knowledge about the molecular structure of GPCRs should pave the way for more efficient drugs. This lab-friendly guide provides a concise, up-to-date presentation of methods which have been used successfully in studying structure and function of GPCRs.
Table of Contents
Partial TOC
Overview of Mutagenesis Techniques .  
The Substituted-Cysteine Accessibility Method  
Metal-Ions as Atomic Scale Probes of G Protein-Coupled Receptor Structure  
Genetic Approaches for Studying the Structure and Function of G Protein-Coupled Receptors in Yeast
Electron-Crystallographic Analysis of Two-Dimensional Rhodopsin Crystals  
Site-Directed Spin-Labeling (SDSL)
Studies of the G Protein-Coupled Receptor Rhodopsin
Lead Discovery and Development for G Protein-Coupled Receptors  
Index.
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?