• USA Home
  • S9764 - Snail acetone powder from Helix pomatia visceral hump

EMAIL THIS PAGE TO A FRIEND
S9764 Sigma

Snail acetone powder from Helix pomatia visceral hump

Purchase

Properties

Related Categories Acetone Powders, Biochemicals and Reagents, Proteins and Derivatives More...
storage temp.   2-8°C

Description

Preparation Note

Acetone powder from visceral hump after removal of most of the albumin glands.

Quality

Contains β-glucuronidase, sulfatase and β-D-mannosidase.

Price and Availability

Safety & Documentation

Safety Information

WGK Germany 
3

Documents

Certificate of Analysis

Protocols & Articles
Related Products

related product

Product #

Description

Add to Cart

G0751 β-Glucuronidase from Helix pomatia, Type H-1, partially purified powder, ≥300,000 units/g solid
S9626 Sulfatase from Helix pomatia, Type H-1, sulfatase ≥10,000 units/g solid

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?