EMAIL THIS PAGE TO A FRIEND
S9813 Sigma
SimVector
Purchase
Properties
Related Categories Computer Software, Labware, Life Science software More...
Description
General description
An exceptional software to design cloning experiments and drawing publication quality plasmid vector maps
• Web integration - Automatically connects to Entrez for sequence retrieval
• Graphical Entrez features - Transforms GenBank annotations to graphical features
• Web ready images - Outputs images in JPEG format for addition to web sites
• Publication quality images - Outputs images in high resolution TIFF format for publication
• Plan cloning experiments - Designs PCR TA cloning, restriction enzyme, and Cre recombinase cloning experiments
• Sequence editing - Easily place inserts in vectors by cutting and pasting
• Active analysis - Automatically updates enzymes and features as a DNA sequence is edited
• Easy styling - Hundreds of fonts, colors, styles and fill patterns for enzymes and features
• Marquee select - Group select objects to quickly set styles for all
• Automatic overlap avoidance - Built in algorithm keeps enzymes and feature labels from overlapping
• Linear and circular views - Produces both circular and linear graphical maps
• Zooming - Zoom in for detailed graphics or out for an overview
• Folders - Keep all sequence files organized by lab, researcher, or project
• Sequence import - Move sequence files from computer to computer or e-mail over the Internet.
• Comprehensive database - Analyzes for restriction sites with database of more than 900 enzymes
• Enzyme Filtering - Select restriction enzymes by overhang, recognition site length, etc.
·
System Requirements
Windows NT / 98 / 2000 / ME Mac OS X
Price and Availability
Safety & Documentation
Safety Information
Safety Information for this product is unavailable at this time.
Documents
Certificate of Analysis
Protocols & Articles
Related Products
Technical Service:
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Bulk Ordering & Pricing:
Need larger quantities for your development, manufacturing or research applications?