• USA Home
  • SAB1101113 - Anti-GATM (101-115) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1101113 Sigma

Anti-GATM (101-115) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-AGAT, Anti-AT, Anti-Glycine amidinotransferase (L-arginine:glycine amidinotransferase)

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, G-GC, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~48 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GATM(2628)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 101-115 of human GATM

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Biochem/physiol Actions

The zinc finger nuclease (ZFN) provided in this kit targets the gene encoding glycine amidinotransferase (GATM). GATM is involved in the formation of ornithine and guanidinoacetate from arginine and glycine. This is an important step in creatine synthesis. Mutations in the gene encoding GATM have been shown to lead to arginine-glycine amidinotransferase deficiency.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?