• USA Home
  • SAB1101559 - Anti-IFIT5 (301-315) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1101559 Sigma

Anti-IFIT5 (301-315) antibody produced in rabbit

IgG fraction of antiserum

Synonym: Anti-Interferon-induced protein with tetratricopeptide repeats 5, Anti-RI58

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, I-IG, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   mol wt ~56 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... IFIT5(24138)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 301-315 of human IFIT5

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?