• USA Home
  • SAB1101764 - Anti-NAT9 (21-35) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1101764 Sigma

Anti-NAT9 (21-35) antibody produced in rabbit

IgG fraction of antiserum

Synonym: Anti-N-acetyltransferase 9 (GCN5-related, putative), Anti-EBSP

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, N-ND, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   mol wt ~23 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... NAT9(26151)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 21-35 of human NAT9

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Application

Anti-NAT9 (21-35) antibody produced in rabbit is suitable for western blot application at a working dilutions of 1:500-1:2,000.

Biochem/physiol Actions

NAT9 (N-acetyltransferase 9) is a member of the N-acetyltransferase family mapped on chromosome 17q25. It has been reported that NAT9 gene may be associated with inflammatory arthritis, referred to as psoriatic arthritis (PsA). Single-nucleotide polymorphisms (SNPs) in NAT9 have been linked to a chronic inflammatory skin disorder, Psoriasis. It has also been reported that RUNX1-mediated defective regulation of NAT9 leads to a susceptible factor responsible for psoriasis.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?