• USA Home
  • SAB1102854 - Anti-PLXNA4 (266-280) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1102854 Sigma

Anti-PLXNA4 (266-280) antibody produced in rabbit

IgG fraction of antiserum

Synonym: Anti-FAYV2820, Anti-KIAA1550, Anti-PLEXA4, Anti-PLXNA4A, Anti-PLXNA4B, Anti-Plexin A4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PJ-PN, Primary Antibodies
species reactivity   human
application(s)   western blot: 1:500-1:2,000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   antigen mol wt ~58 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... PLXNA4(91584)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to amino acids 266-280 of human PLXNA4

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?