• USA Home
  • SAB1303862 - ANTI-ZNF354A (N-TERM) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1303862 Sigma

ANTI-ZNF354A (N-TERM) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: EZNF, HKL1, TCF17, Transcription factor 17, ZNF354A, Zinc finger protein 354A, Zinc finger protein eZNF

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, ZNF0-ZNF3
species reactivity   human
application(s)   flow cytometry: 1:10-1:50
  western blot: 1:250-1:500
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   mol wt 69237 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... (6940)
UniProt accession no.   O60765

Description

Physical form

Purified polyclonal antibody supplied in PBS with 0.09% (W/V) sodium azide. This antibody is prepared by Saturated Ammonium Sulfate (SAS) precipitation followed by dialysis against PBS.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?