• USA Home
  • SAB1306340 - ANTI-PAI2 (CENTER) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1306340 Sigma

ANTI-PAI2 (CENTER) antibody produced in rabbit

purified immunoglobulin, buffered aqueous solution

Synonym: Monocyte Arg-serpin, PAI2, PLANH2, Plasminogen activator inhibitor 2, SERPINB2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, P-PA, Primary Antibodies
species reactivity   human
application(s)   immunohistochemistry: 1:10-1:50
  western blot: 1:1000
clone   polyclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
mol wt   mol wt 46596 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... SERPINB2(5055)
biological source   rabbit
NCBI accession no.   NP_001137290.1
UniProt accession no.   P05120

Description

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?