• USA Home
  • SAB1306352 - ANTI-PKC GAMMA(C-TERMINAL) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1306352 Sigma

ANTI-PKC GAMMA(C-TERMINAL) antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: PKCG, PRKCG, Protein kinase C gamma type

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PJ-PN, Primary Antibodies
species reactivity   human, mouse
application(s)   immunohistochemistry: 1:10-1:50
  western blot: 1:1000
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
mol wt   mol wt 78448 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... PRKCG(5582)
biological source   rabbit
NCBI accession no.   NP_002730.1
UniProt accession no.   P05129

Description

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Articles

Chondroitinase for Neural Regeneration Research

The spinal cord is the information highway system for the brain. And axons are the road that allows information data to be transferred. So, when a traumatic spinal cord injury (SCI) occurs and axons ...
Keywords: Catalysis, Cell culture, Degradations, High performance liquid chromatography, Immunofluorescence, PAGE

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?