• USA Home
  • SAB1306399 - ANTI-ASK1 (MEKK5/MAP3K5) (S83) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB1306399 Sigma

ANTI-ASK1 (MEKK5/MAP3K5) (S83) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: ASK1, MAP3K5, MAPKKK5, MEKK5, Mitogen-activated protein kinase kinase kinase 5

Purchase

Properties

Related Categories AS-ATG, Alphabetical Index, Antibodies, Primary Antibodies
species reactivity   human
application(s)   immunohistochemistry: 1:10-1:50
  western blot: 1:1000
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   mol wt 154537 Da
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... MAP3K5(4217)
biological source   rabbit
NCBI accession no.   NP_005914.1
UniProt accession no.   Q99683

Description

Physical form

Supplied in PBS with 0.09% (W/V) sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?