• USA Home
  • SAB1400441 - Anti-SNX13 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB1400441 Sigma

Anti-SNX13 antibody produced in mouse

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-KIAA0713, Anti-RGS-PX1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, SM-SN
species reactivity   human
application(s)   capture ELISA: suitable
  indirect immunofluorescence: suitable
  western blot: suitable
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... SNX13(23161)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

SNX13 (AAH45667.1, 1 a.a. ~ 957 a.a) full-length human protein.

Sequence
MLTEASLSIWGWGSLGIVLFLITFGPFVIFYLTFYILCFVGGGLVVTLLFGKTNSEKYLEQCEHSFLPPTSPGVPKCLEEMKREARTIKIDRRLTGANIIDEPLQQVIQFSLRDYVQYWYYTLSDDESFLLEIRQTLQNALIQFATRSKEIDWQPYFTTRIVDDFGTHLRVFRKAQQKITEKDDQVKGTAEDLVDTFFEVEVEMEKEVCRDLVCTSPKDEEGFLRDLCEVLLYLLLPPGDFQNKIMRYFVREILARGILLPLINQLSDPDYINQYVIWMIRDSNCNYEAFMNIIKLSDNIGELEAVRDKAAEELQYLRSLDTAGDDINTIKNQINSLLFVKKVCDSRIQRLQSGKEINTVKLAANFGKLCTVPLDSILVDNVALQFFMDYMQQTGGQAHLFFWMTVEGYRVTAQQQLEVLLSRQRDGKHQTNQTKGLLRAAAVGIYEQYLSEKASPRVTVDDYLVAKLADTLNHEDPTPEIFDDIQRKVYELMLRDERFYPSFRQNALYVRMLAELDMLKDPSFRGSDDGDGESFNGSPTGSINLSLDDLSNVSSDDSVQLHAYISDTGVCNDHGKTYALYAITVHRRNLNSEEMWKTYRRYSDFHDFHMRITEQFESLSSILKLPGKKTFNNMDRDFLEKRKKDLNAYLQLLLAPEMMKASPALAHYVYDFLENKAYSKGKGDFARKMDTFVNPLRNSMRNVSNAVKSLPDNLAEGMTKMSDNMGKMSERLGQDIKQSFFKVPPLIPKTDSDPEHRRVSAQLDDNVDDNIPLRVMLLLMDEVFDLKERNQWLRRNIKNLLQQLIRATYGDTINRKIVDHVDWMTSPEQVADSVKRFRDAFWPNGILAEAVPCRDKSIRMRTRVAGKTKLLAIMPDELKHIIGAETTRKGILRVFEMFQHNQLNRRMVYVFLEGFLETLFPQYKFRELFNKLHSRSKQMQKYKQKLQTTQAPSLQKR

General description

This gene encodes a PHOX domain- and RGS domain-containing protein that belongs to the sorting nexin (SNX) family and the regulator of G protein signaling (RGS) family. The PHOX domain is a phosphoinositide binding domain, and the SNX family members are involved in intracellular trafficking. The RGS family members are regulatory molecules that act as GTPase activating proteins for G alpha subunits of heterotrimeric G proteins. The RGS domain of this protein interacts with G alpha(s), accelerates its GTP hydrolysis, and attenuates G alpha(s)-mediated signaling. Overexpression of this protein delayes lysosomal degradation of the epidermal growth factor receptor. Because of its bifunctional role, this protein may link heterotrimeric G protein signaling and vesicular trafficking. (provided by RefSeq)

Physical form

Solution in phosphate buffered saline, pH 7.4

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?