| Related Categories | Alphabetical Index, Antibodies, MF-MN, Primary Antibodies |
| species reactivity | human |
| application(s) | indirect immunofluorescence: suitable |
| western blot: suitable | |
| concentration | ~0.45 mg/mL |
| antibody form | purified immunoglobulin |
| form | buffered aqueous solution |
| immunogen sequence |
MDKDCEMKRTTLDSPLGKLELSGCE |
| mol wt | antigen mol wt ~21.6 kDa |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... MGMT(4255) |
| NCBI accession no. |
NM_002412.2 |
MGMT (NP_002403.1, 1 a.a. ~ 207 a.a) full-length human protein.
Mouse polyclonal antibody raised against a full-length human MGMT protein.
Clear, colorless solution in phosphate buffered saline, pH 7.2
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Customers Also Viewed
clone MGMT-214, purified immunoglobulin, buffered aqueous solution
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?