Synonym: Anti-H2A histone family, member X, Anti-H2A.X, Anti-H2AX
MDL number MFCD07370338
| Related Categories | Alphabetical Index, Antibodies, Antibodies to Histones, Epigenetics, H1-HE, |
| species reactivity | human, mouse, rat, bovine, pig, zebrafish, canine |
| application(s) | western blot: suitable |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | lyophilized powder |
| immunogen sequence |
SGRGKTGGKARAKAKSRSSRAGLQF |
| storage temp. | −20°C |
| Gene Information | human ... H2AFX(3014) |
Peptide region of the protein sequence according to NP_002096.
Lyophilized from PBS buffer with 2% sucrose
Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.
Customers Also Viewed
clone 3F4, purified immunoglobulin, buffered aqueous solution
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?