• USA Home
  • SAB2103305 - Anti-DDAH2, (N-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2103305 Sigma

Anti-DDAH2, (N-terminal) antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym: Anti-DDAH, Anti-DDAHII, Anti-G6a, Anti-NG30

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, DA-DD, Primary Antibodies
species reactivity   rat, mouse, horse, human, rabbit, bovine
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
immunogen sequence   MYTLHTKRVKAAARQMWTSNLSKVRQSLKNVYHKCKIRHQDSTGYPTVTS
mol wt   mol wt 31 kDa
storage temp.   −20°C
Gene Information   human ... DDAH2(23564)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   NM_013974

Description

Immunogen

Peptide region of the protein sequence according to NP_039268.

Physical form

Antibody is lyophilized from PBS buffer with 2% sucrose.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?