• USA Home
  • SAB2104109 - Anti-PAFAH1B1, (N-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2104109 Sigma

Anti-PAFAH1B1, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym: Anti-LIS1, Anti-LIS2, Anti-MDCR, Anti-MDS, Anti-PAFAH

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, P-PA, Primary Antibodies
species reactivity   canine, zebrafish, human, mouse, bovine, horse, guinea pig, rabbit, pig, rat
application(s)   western blot: suitable
clone   polyclonal
concentration   0.5 mg - 1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   mol wt 47 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... PAFAH1B1(5048)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   NM_000430

Description

Immunogen

The immunogen for anti-PAFAH1B1 antibody: synthetic peptide derected towards the N terminal of human PAFAH1B1

General description

PAFAH1B1 (Platelet activating factor acetylhydrolase 1b regulatory subunit 1) encodes a protein referred to as Lis1. It is located on the chromosome location 17p13.3. The gene spans ~92 kb at the genomic level. It consists of a N-terminal coiled-coil domain and seven WD40 repeats at the C-terminus end.

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Application

Anti-PAFAH1B1, (N-terminal) antibody produced in rabbit is suitable for western blot analysis.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Biochem/physiol Actions

PAFAH1B1 (Platelet activating factor acetyl hydrolase 1b regulatory subunit 1) majorly participates in the neuronal migration pathway during brain development. It is a non-catalytic regulatory subunit of platelet activating factor (PAF) acetyl hydrolase 1b (Pafah1b) complex. During neuronal migration, it conjugates with PAFAH1B2 and PAFAH1B3 to control the concentration of platelet activating factor in the brain. Deleted gene expression of PAFAH1B1 has been reported in the 7p13.3 deletion syndrome or Miller-Dieker syndrome with mental retardation and facial dysmorphism. Heterozygous mutations of the gene cause type 1 lissencephaly characterized with impaired neuronal migration and reduction in the number of cortical gyri.

Sequence

Synthetic peptide located within the following region: MVLSQRQRDELNRAIADYLRSNGYEEAYSVFKKEAELDVNEELDKKYAGL

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?