• USA Home
  • SAB2104155 - Anti-PRDM11 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2104155 Sigma

Anti-PRDM11 antibody produced in rabbit

affinity isolated antibody, lyophilized powder

Synonym: Anti-PFM8

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PR-PR, Primary Antibodies
species reactivity   horse, mouse, human, rabbit, rat
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
immunogen sequence   PGEWLRVWYSEDYMKRLHSMSQETIHRNLARGEKRLQREKSEQVLDNPED
mol wt   mol wt 58 kDa
storage temp.   −20°C
Gene Information   human ... PRDM11(56981)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   NM_020229

Description

Immunogen

Peptide region of the protein sequence according to NP_064614.

Physical form

Antibody is lyophilized from PBS buffer with 2% sucrose.

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?