• USA Home
  • SAB2104215 - Anti-ACCN2, (N-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2104215 Sigma

Anti-ACCN2, (N-terminal) antibody produced in rabbit

affinity isolated antibody

Synonym: Anti-ASIC1A, Anti-ASIC, Anti-BNaC2, Anti-hBNaC2

Purchase

Properties

Related Categories AC-AC, Alphabetical Index, Antibodies, Primary Antibodies
species reactivity   rabbit, mouse, horse, human, rat, guinea pig
application(s)   western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   lyophilized powder
mol wt   mol wt 65 kDa
storage temp.   −20°C
Gene Information   human ... ACCN2(41)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   NM_020039

Description

Immunogen

The immunogen for anti-ACCN2 antibody: synthetic peptide derected towards the N terminal of human ACCN2

Physical form

Lyophilized from PBS buffer with 2% sucrose

Features and Benefits

Antibody Bioguarantee
Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Sequence

Synthetic peptide located within the following region: MELKAEEEEVGGVQPVSIQAFASSSTLHGLAHIFSYERLSLKRALWALCF

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?