• USA Home
  • SAB2500076 - Anti-APH1A antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
SAB2500076 Sigma

Anti-APH1A antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-6530402N02Rik, Anti-Anterior pharynx defective 1 homolog A, Anti-CGI-78

Purchase

Properties

Related Categories AO-AQ, Alphabetical Index, Antibodies, Antibodies for Alzheimer′s Disease, Antibodies for Neurobiology,
species reactivity   rat, human, mouse
application(s)   indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... APH1A(51107)
biological source   goat
conjugate   unconjugated

Description

Immunogen

Peptide with sequence C-HVTDRSDARLQYG from the internal region of the protein sequence according to NP_001071096.1; NP_057106.2.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Symbol  GHS07  GHS07
Signal word  Warning
Hazard statements  H315-H319
Precautionary statements  P305 + P351 + P338
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Articles

Alzheimer's Disease

Alzheimer's disease (AD) is the most common cause of dementia in the elderly and is characterized by gradual loss of cognitive functions. Hallmark pathohistological findings of AD include widespread ...
Carolyn L. Crankshaw
BioFiles v7 n2, 2011, 4–8
Keywords: Alzheimer Disease, Gene expression, Genetic, Genetics, Inflammation, Metabolism, Phosphorylations

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?