• USA Home
  • SAB2500168 - Anti-BMYB/MYBL2 antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
SAB2500168 Sigma

Anti-BMYB/MYBL2 antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-HGNC:7548, Anti-MGC15600, Anti-MYB-related protein B, Anti-MYBL2, Anti-v-myb Myeloblastosis viral oncogene homolog (avian)-like 2

Purchase

Properties

Related Categories AA - CA, Alphabetical Index, Antibodies, Antibodies to Transcription Factors, BL-BZ,
species reactivity   mouse, canine, rat, human
application(s)   indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... MYBL2(4605)
biological source   goat
conjugate   unconjugated

Description

Immunogen

Peptide with sequence C-QEKARQLLGRLKPSH from the C Terminus of the protein sequence according to NP_002457.1.

Physical form

Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Symbol  GHS07  GHS07
Signal word  Warning
Hazard statements  H315-H319
Precautionary statements  P305 + P351 + P338
RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?