Synonym: Anti-DKFZp434H2111, Anti-FLJ45829, Anti-Leucine-rich repeat kinase 2, Anti-PARK8, Anti-ROCO2
| Related Categories | Alphabetical Index, Antibodies, Antibodies for Parkinson’s Disease Research, Cell Biology, Cell Signaling and Neuroscience, |
| species reactivity | human |
| application(s) | immunohistochemistry: suitable |
| indirect ELISA: suitable | |
| western blot: suitable | |
| clone | polyclonal |
| antibody form | affinity isolated antibody |
| form | buffered aqueous solution |
| shipped in | dry ice |
| storage temp. | −20°C |
| Gene Information | human ... LRRK2(120892) |
Peptide with sequence CELAEKMRRTSV from the internal region (near the C Terminus) of the protein sequence according to NP_940980.2.
Supplied at 0.5 mg/mL in Tris saline with 0.02% sodium azide and 0.5% bovine serum albumin.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Need larger quantities for your development, manufacturing or research applications?