• USA Home
  • SAB2501411 - Anti-GNAS antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
SAB2501411 Sigma

Anti-GNAS antibody produced in goat

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-AHO, Anti-C20orf45, Anti-GNAS1, Anti-GPSA, GNAS complex locus

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, GM-GO, Primary Antibodies
species reactivity   bovine, mouse, rat, bovine, human
application(s)   indirect ELISA: suitable
  indirect immunofluorescence: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GNAS(2778)
biological source   goat
conjugate   unconjugated

Description

Immunogen

Peptide with sequence QAARSNSDGEKATK, from the internal region of the protein sequence according to NP_000507.1; NP_001070956.1; NP_001070957.1

Physical form

Supplied at 0.5 mg/mL in 20mM Tris (pH 7.3) and 150mM NaCl with 0.02% sodium azide and 0.5% bovine serum albumin.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?