• USA Home
  • SAB2900008 - Anti-TRH-R antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2900008 Sigma

Anti-TRH-R antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: thyroliberin endocrine receptor, thyroliberin receptor, thyrotropin-releasing factor receptor, thyrotropin-releasing hormone receptor, trh receptor

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, TR-TR
species reactivity   sheep, pig, human, horse, dog
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... TRHR(7201)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

Synthetic 16 amino acid peptide from C-terminus of human TRH Receptor. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Sheep, Dog, Elephant, Panda, Horse, Pig (100%); Marmoset, Mouse, Rat, Bovine, Hamster, Opossum, Turkey, Chicken, Lizard, Xenopus (94%); Stickleback (88%); Rabbit, Zebrafish (81%).

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?