• USA Home
  • SAB2900109 - Anti-OGR1/GPR68 (AB1) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2900109 Sigma

Anti-OGR1/GPR68 (AB1) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: GPR68, brgr1, brgrb, g protein-coupled receptor 68, g protein-coupled receptor ogr1, gpr12a

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, O, Primary Antibodies
species reactivity   monkey, rat, human
application(s)   ELISA: suitable
  immunohistochemistry (formalin-fixed, paraffin-embedded sections): 10-20 μg/mL
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... GPR68(8111)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

Synthetic 18 amino acid peptide from 1st extracellular domain of human GPR68. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Rat (100%); Mouse, Bat, Dog, Hamster, Panda, Horse, Pig (94%); Bovine, Rabbit, Platypus (89%); Elephant (83%).

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?