• USA Home
  • SAB2900233 - Anti-ITGA2 (AB2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2900233 Sigma

Anti-ITGA2 (AB2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: alpha2 integrin, cd49b, integrin alpha 2, platelet membrane glycoprotein ia

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, IQ-IZ, Primary Antibodies
species reactivity   pig, human, monkey, dog
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ITGA2(3673)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

Synthetic 16 amino acid peptide from extracellular domain of human ITGA2 / CD49b. Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Monkey, Marmoset, Dog, Panda, Pig (100%); Bovine, Elephant, Horse, Chicken, Lizard (94%); Opossum, Turkey (88%); Bat, Hamster, Stickleback (81%).

Physical form

Solution in phosphate-buffered saline containing less than 0.1% sodium azide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?