• USA Home
  • SAB2900329 - Anti-DDB1 (internal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2900329 Sigma

Anti-DDB1 (internal) antibody produced in rabbit

whole antiserum

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, DA-DD, Primary Antibodies, TR-TR More...
species reactivity   human
application(s)   ELISA: suitable
  immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  immunoprecipitation: suitable
  western blot: suitable
clone   polyclonal
antibody form   whole antiserum
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... DDB1(1642)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

Synthetic peptide from human DDB1.

Physical form

0.01% sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?