• USA Home
  • SAB2900337 - Anti-Fibrinogen (AB1) antibody produced in goat

EMAIL THIS PAGE TO A FRIEND
SAB2900337 Sigma

Anti-Fibrinogen (AB1) antibody produced in goat

fractionated antiserum, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, FB-FJ, Primary Antibodies
biological source   goat
antibody form   fractionated antiserum
clone   polyclonal
form   buffered aqueous solution
species reactivity   human
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  immunoprecipitation: suitable
  indirect immunofluorescence: suitable
conjugate   unconjugated
shipped in   dry ice
storage temp.   −20°C

Description

Immunogen

Purified fibrinogen from human plasma.

Physical form

0.02 M potassium phosphate, 0.15 M sodium chloride, pH 7.2, 0.01% sodium azide

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

Safety Information for this product is unavailable at this time.
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?