• USA Home
  • SAB2900386 - Anti-IRF7 (AB2) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB2900386 Sigma

Anti-IRF7 (AB2) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Transcription Factors, Epigenetics, HO - L999,
species reactivity   mouse, human, rat
application(s)   immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... IRF7(3665)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

Synthetic peptide from human IRF7.

Physical form

Solution in phosphate-buffered saline containing 0.02% sodium azide.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?