• USA Home
  • SAB3500118 - Anti-OCIAD2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3500118 Sigma

Anti-OCIAD2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution, ab1

Synonym: Anti-OCIA domain-containing protein 2, Anti-Ovarian cancer immuno-reactive antigen domain-containing protein 2

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, N-P, O,
species reactivity   human
application(s)   immunofluorescence: suitable
  indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... OCIAD2(132299)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   NP_001014446
UniProt accession no.   Q56VL3

Description

Immunogen

OCIAD2 antibody was raised against an 18 amino acid peptide from near the carboxy terminus of human OCIAD2.

Antigen Background

OCIAD2 was identified by its sequence similarity with OCIAD1, and together OCIAD1 and OCIAD2 form the OCIA domain family. OCIAD2 mRNA was found to be expressed at higher levels in invasive adenocarcinoma mixed subtype with bronchioloalveolar carcinoma component (BAC) of the lung. Loss of OCIAD2 expression was significantly correlated with lymphatic invasion, blood vessel invasion, and lymph node metastasis, indicating that OCIAD2 may play a role in cell adhesion and prevention of cell migration. While the function of OCIAD2 is still unknown, its expression in adenocarcinoma with BAC component is significantly associated with a favorable prognosis and may serve as a marker for selecting tumors that are treatable by limited surgery.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500118.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

related product

Product #

Description

Add to Cart

SBP3500118 OCIAD2 (AB1) Blocking Peptide for SAB3500118

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?