• USA Home
  • SAB3500129 - Monoclonal Anti-IRAK antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB3500129 Sigma

Monoclonal Anti-IRAK antibody produced in mouse

clone 8F1A7, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-IL-1-Receptor Associated Kinase

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, IQ-IZ, Primary Antibodies
biological source   mouse
antibody form   purified immunoglobulin
clone   8F1A7, monoclonal
form   buffered aqueous solution
species reactivity   mouse, human
application(s)   immunohistochemistry: suitable
  indirect ELISA: suitable
  western blot: suitable
conjugate   unconjugated
UniProt accession no.   P51617
shipped in   dry ice
storage temp.   −20°C

Description

Antigen Background

Nuclear factor kappa B (NF-kappaB) is a ubiquitous transcription factor and an essential mediator of gene expression during activation of immune and inflammatory responses. NF-kappaB mediates the expression of a great variety of genes in response to extracellular stimuli including IL-1, TNFα and LPS. A serine/threonine protein kinase associated with IL-1 receptor (IRAK) and its homologue mouse pelle-like protein kinase (mPLK) were identified recently. IRAK is associated with the IL-1 receptor subunits IL-1RI and IL-1RAcP after IL-1 binding and serves as a signaling molecule to mediate IL-1 response. IRAK mediates a signaling cascade leading to NF-kB activation by members in IL-1 family including IL-1 and a novel cytokine IL-18 (also termed IGIF).

Immunogen

raised against a recombinant protein corresponding to amino acids 618 - 712 of human IRAK.

Physical form

Supplied at approx. 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR 
NONH for all modes of transport
WGK Germany 
2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?