• USA Home
  • SAB3500131 - Anti-VLK antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3500131 Sigma

Anti-VLK antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-PKDCC, Anti-Protein kinase domain containing cytoplasmic homolog, Anti-SGK439, Anti-Vertebrate lonesome kinase

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, Primary Antibodies, U-Z,
species reactivity   mouse, human, rat
application(s)   immunofluorescence: suitable
  immunohistochemistry: suitable
  indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   predicted mol wt 32-54 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... Pkdcc(106522)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   BAH66379
UniProt accession no.   Q5RJI4

Description

Immunogen

VLK antibody was raised against a 15 amino acid peptide from near the center of human VLK.

General description

Protein kinase domain containing, cytoplasmic homolog (PKDCC) is a secreted protein expressed in platelets. The gene encoding it is localized on human chromosome 2p21.

Physical form

Supplied in PBS with 0.02% sodium azide.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500131.

Biochem/physiol Actions

Protein kinase domain containing, cytoplasmic homolog (PKDCC) is crucial for embryonic development. It phosphorylates proteins associated with the endoplasmic reticulum and other secreted proteins. PKDCC has been linked to atopy.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

SBP3500131 VLK Blocking Peptide for SAB3500131

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?