• USA Home
  • SAB3500147 - Anti-ATM antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3500147 Sigma

Anti-ATM antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-Mutated in Ataxia-Telangiectasia

Purchase

Properties

Related Categories A-B, ATH-AZ, Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides,
species reactivity   human
application(s)   immunofluorescence: suitable
  immunohistochemistry: suitable
  indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... ATM(472)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   AAB65827
UniProt accession no.   Q13315

Description

Immunogen

ATM antibody was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human ATM.

Physical form

Supplied in PBS with 0.02% sodium azide.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500147.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

SBP3500147 ATM Blocking Peptide for SAB3500147

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?