• USA Home
  • SAB3500189 - Anti-Bonzo antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3500189 Sigma

Anti-Bonzo antibody produced in rabbit

IgG fraction of antiserum, buffered aqueous solution

Synonym: Anti-STRL33

Purchase

Properties

Related Categories A-B, Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, BL-BZ,
species reactivity   human
application(s)   indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   IgG fraction of antiserum
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... CXCR6(10663)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   AAB64221
UniProt accession no.   O00574

Description

Immunogen

Bonzo antibody was raised against a peptide corresponding to amino acids 319 to 338 of human origin. The sequence of this peptide is identical to those of macaque and African green monkey.

Physical form

Supplied in PBS with 0.02% sodium azide.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500189.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

related product

Product #

Description

Add to Cart

SBP3500189 Bonzo Blocking Peptide for SAB3500189

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?