• USA Home
  • SAB3500219 - Anti-Fibulin 3 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3500219 Sigma

Anti-Fibulin 3 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym: Anti-DHRD, Anti-DRAD, Anti-EFEMP1, Anti-EGF-containing fibulin-like extracellular matrix protein 1, Anti-FBLN3

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, F-J, FB-FJ,
species reactivity   human, rat, mouse
application(s)   immunocytochemistry: suitable
  immunofluorescence: suitable
  indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   predicted mol wt 52 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... EFEMP1(2202)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   NP_004096
UniProt accession no.   Q12805

Description

Immunogen

Fibulin 3 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human Fibulin 3.

Physical form

Supplied in PBS with 0.02% sodium azide.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500219.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

related product

Product #

Description

Add to Cart

SBP3500219 Fibulin 3 Blocking Peptide for SAB3500219

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?