• USA Home
  • SAB3500239 - Anti-DAPK2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB3500239 Sigma

Anti-DAPK2 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies with Corresponding Blocking Peptides, C-E, DA-DD,
species reactivity   mouse, rat, human
application(s)   immunohistochemistry: suitable
  indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   predicted mol wt 42 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... DAPK2(23604)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   BAA88063
UniProt accession no.   Q9UIK4

Description

Immunogen

DAPK2 antibody was raised against a peptide corresponding to amino acids near the carboxy terminus of human DAPK2. The sequence of this antigenic peptide is identical to the corresponding amino acids of mouse origin (1,2).

Physical form

Supplied in PBS with 0.02% sodium azide.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500239.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

related product

Product #

Description

Add to Cart

SBP3500239 DAP kinase 2 Blocking Peptide for SAB3500239

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?