• USA Home
  • SAB3500467 - Monoclonal Anti-PUMA antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB3500467 Sigma

Monoclonal Anti-PUMA antibody produced in mouse

clone 10D4G7, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-Bbc3, Anti-Bcl-2 binding component 3, Anti-P53 upregulated modulator of apoptosis

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, PT-PY, Primary Antibodies
species reactivity   human, rat
application(s)   indirect ELISA: suitable
  western blot: suitable
clone   10D4G7, monoclonal
antibody form   purified immunoglobulin
form   buffered aqueous solution
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... BBC3(27113)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   Q9BXH1
UniProt accession no.   Q9BXH1

Description

Immunogen

Mouse monoclonal PUMA antibody was raised against a recombinant protein corresponding to amino acids 76 – 170 of human PUMA-alpha.

Physical form

Supplied in PBS with 0.02% sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?