• USA Home
  • SAB4100053 - Monoclonal Anti-NME4 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4100053 Sigma

Monoclonal Anti-NME4 antibody produced in mouse

clone NME4-C17, culture supernatant

Synonym: Anti-NDPK-D, Anti-NM23H4, Anti-Non-metastatic cells 4, Anti-nm23-H4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, NK-NR, Primary Antibodies
application(s)   western blot: suitable
clone   NME4-C17, monoclonal
antibody form   culture supernatant
form   aqueous solution
isotype   IgG1
mol wt   antigen predicted mol wt 20.6 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... NME4(4833)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

NME4 purified recombinant protein

Physical form

Solution in Dulbecco′s Modified Eagle′s Medium with 10% fetal calf serum and 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  3
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?