• USA Home
  • SAB4200253 - Monoclonal Anti-triMethyl-Histone H4 (triMe-Lys20) antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4200253 Sigma

Monoclonal Anti-triMethyl-Histone H4 (triMe-Lys20) antibody produced in mouse

~1.0 mg/mL, clone 6F8-D9, purified immunoglobulin

Synonym: Anti-H4K20me3, Anti-HIST4H4

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies to Histones, Epigenetics, Primary Antibodies,
species reactivity   human, mouse, rat
application(s)   indirect immunofluorescence: 5-10 μg/mL using cultured human HeLa cells
  western blot: 1-2 μg/mL using histones isolated from human HeLa cells
clone   6F8-D9, monoclonal
concentration   ~1.0 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   mol wt ~14 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... HIST4H4(121504)
mouse ... Hist1h4(319156)
rat ... Hist2h4(295277)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

hybridoma 6F8-D9 produced by the fusion of mouse myeloma cells and splenocytes from BALB/c mice immunized with a peptide containing trimethylated Lys20 of human Histone H4

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Inflammation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?