• USA Home
  • SAB4200373 - Anti-WASH1 (C-terminal) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4200373 Sigma

Anti-WASH1 (C-terminal) antibody produced in rabbit

~1.0 mg/mL, affinity isolated antibody

Synonym: Anti-FAM39E, Anti-WAS protein family homolog 1, Anti-WASH

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies to Actin-associated Proteins/Myosin, Antibodies to Cell and Organelle Proteins,
species reactivity   human, rat, mouse
application(s)   indirect immunofluorescence: 2.5-5.0 μg/mL using rat NRK cells.
  western blot: 2-4 μg/mL using whole extracts of human HeLa / K562 and mouse RAW-264 / A9 cells.
clone   polyclonal
concentration   ~1.0 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt ~72 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... WASH1(100287171)
mouse ... ORF19(68767)
rat ... Wash2(367328)
biological source   rabbit
conjugate   unconjugated

Description

Immunogen

synthetic peptide corresponding to the C-terminal region of human WASH1, conjugated to KLH. The corresponding sequence is identical in mouse and rat.

Physical form

Solution in 0.01 M phosphate buffered saline pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?