• USA Home
  • SAB4200445 - Monoclonal Anti-LRIG1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4200445 Sigma

Monoclonal Anti-LRIG1 antibody produced in mouse

~1.0 mg/mL, clone LRIG1-21, purified immunoglobulin

Synonym: Anti-LIG1, Anti-Leucine-rich repeats and immunoglobulin-like domains 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Cell Stress, Antibodies for Nitric Oxide Research,
species reactivity   human
application(s)   immunocytochemistry: suitable
  immunohistochemistry: suitable
  indirect immunofluorescence: 2.5-5.0 μg/mL using LNCaP cells
  western blot: 2-4 μg/mL using extracts of LNCaP cells
clone   LRIG1-21, monoclonal
concentration   ~1.0 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
mol wt   antigen mol wt ~110 kDa
  antigen mol wt ~143 kDa
shipped in   dry ice
storage temp.   −20°C
Gene Information   human ... LRIG1(26018)
biological source   mouse
conjugate   unconjugated

Description

Immunogen

peptide corresponding to a sequence at the N-terminus of human LRIG1, conjugated to KLH.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  2
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?