• USA Home
  • SAB4503507 - Anti-RAB7L1 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4503507 Sigma

Anti-RAB7L1 antibody produced in rabbit

affinity isolated antibody

Synonym: RAB7, RAB7L, member RAS oncogene family-like 1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, RAB-RAD
species reactivity   human, mouse, rat
application(s)   indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 23 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... RAB7L(8934)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   O14966

Description

Immunogen

Synthesized peptide derived from human RAB7L1

General description

RAB7L1 (Rab-7-like protein 1, also referred to as RAB29) is a cytosolic GTPase. The gene encoding it is localized on human chromosome 1q32. Anti-RAB7L1 Antibody detects endogenous levels of total RAB7L1 protein.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Biochem/physiol Actions

RAB7L1 (Rab-7-like protein 1, also referred to as RAB29) modulates protein transport from the Golgi apparatus to lysosomes. It also modulates phagocytosis. RAB7L1 has been associated with Parkinson’s disease.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?