• USA Home
  • SAB4503519 - Anti-STAM2 antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4503519 Sigma

Anti-STAM2 antibody produced in rabbit

affinity isolated antibody

Synonym: HBP, HRS binding protein, Signal transducing adaptor molecule (SH3 domain and ITAM motif) 2, Signal transducing adaptor molecule 2A, Signal transducing adaptor molecule 2B

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, ST-ST
species reactivity   human, mouse
application(s)   indirect immunofluorescence: suitable
  western blot: suitable
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 58 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... STAM2(10254)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   O75886

Description

Immunogen

Synthesized peptide derived from human STAM2

General description

Anti-STAM2 Antibody detects endogenous levels of total STAM2 protein.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?