• USA Home
  • SAB4503723 - Anti-POM121, C-Terminal antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4503723 Sigma

Anti-POM121, C-Terminal antibody produced in rabbit

affinity isolated antibody

Synonym: Nuclear envelope pore membrane protein POM 121, P145, Pore membrane protein of 121 kDa

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, POD-POZ, Primary Antibodies
species reactivity   mouse, human, rat
application(s)   indirect ELISA: suitable
  western blot: suitable
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 125 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... POM121(9883)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   Q9Y2N3

Description

Immunogen

Synthesized peptide derived from human POM121

General description

Anti-POM121 Antibody detects endogenous levels of total POM121 protein.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  nwg
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?