• USA Home
  • SAB4504081 - Anti-phospho-Bax (pThr167) antibody produced in rabbit

EMAIL THIS PAGE TO A FRIEND
SAB4504081 Sigma

Anti-phospho-Bax (pThr167) antibody produced in rabbit

affinity isolated antibody

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Cell Biology, Antibodies for Kinase/Phosphatase Biology, Antibodies to Phosphoproteins at Phosphothreonine (pThr),
species reactivity   rat, human, mouse
application(s)   immunohistochemistry: suitable
  indirect ELISA: suitable
clone   polyclonal
concentration   ~1 mg/mL
antibody form   affinity isolated antibody
form   buffered aqueous solution
mol wt   antigen mol wt 21 kDa
shipped in   wet ice
storage temp.   −20°C
Gene Information   human ... BAX(581)
biological source   rabbit
conjugate   unconjugated
NCBI accession no.   Q07812

Description

Immunogen

The antiserum was produced against synthesized peptide derived from human Bax around the phosphorylation site of Thr167.

Physical form

Rabbit IgG in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, 0.02% sodium azide and 50% glycerol.

Price and Availability


Your Key to Success

Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
WGK Germany  1
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Peer-Reviewed Papers
15

References

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?