• USA Home
  • SAB4700166 - Monoclonal Anti-CD36 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4700166 Sigma

Monoclonal Anti-CD36 antibody produced in mouse

clone CB38 (NL07), purified immunoglobulin, buffered aqueous solution

Synonym: Anti-GPIIIb

Properties

Related Categories Alphabetical Index, Antibodies, Antibodies for Flow Cytometry, Antibodies to CD and Related Antigens, CD0-CDC,
species reactivity   human
application(s)   flow cytometry: suitable
clone   CB38 (NL07), monoclonal
concentration   1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgM
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... CD36(948)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   NM_000072.3

Description

Immunogen

living human myeloid cells

General description

The mouse monoclonal antibody CB38 (NL07) recognizes CD36 (GPIIIb), a 85-113 kDa integral membrane glycoprotein expressed on platelets, macrophages, endothelial cells, early erythroid cells and megakaryocytes.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Application

The reagent is designed for Flow Cytometry analysis. Suggested working dilution is 1 μg/mL of sample. Indicated dilution is recommended starting point for use of this product. Working concentrations should be determined by the investigator.

Price and Availability


Detect Your Unseen Protein

Flow Cytometry Instruments

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?