• USA Home
  • SAB4700752 - Monoclonal Anti-Ubinuclein 1 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4700752 Sigma

Monoclonal Anti-Ubinuclein 1 antibody produced in mouse

clone UBN1-02, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-UBN1

Purchase

Properties

Related Categories Alphabetical Index, Antibodies, Primary Antibodies, U1-UG
species reactivity   human
application(s)   immunoblotting: suitable
  immunoprecipitation: suitable
clone   UBN1-02, monoclonal
concentration   1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... UBN1(29855)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   NM_001079514.1

Description

Immunogen

Recombinant protein corresponding to amino acids 1-190 of ubinuclein 1.

General description

The antibody UBN1-02 recognizes N-terminal part of ubinuclein 1 (UBN1), a widely expressed nuclear and adhesion complex protein. Western blotting analysis reveals UBN1 as a 165 kDa band.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?