• USA Home
  • SAB4700753 - Monoclonal Anti-AGR3 antibody produced in mouse

EMAIL THIS PAGE TO A FRIEND
SAB4700753 Sigma

Monoclonal Anti-AGR3 antibody produced in mouse

clone AGR3.2, purified immunoglobulin, buffered aqueous solution

Synonym: Anti-AG3, Anti-hAG-3

Purchase

Properties

Related Categories AE-AK, Alphabetical Index, Antibodies, Primary Antibodies
species reactivity   human
application(s)   immunoblotting: suitable
  immunocytochemistry: suitable
  immunohistochemistry: suitable
clone   AGR3.2, monoclonal
concentration   1 mg/mL
antibody form   purified immunoglobulin
form   buffered aqueous solution
isotype   IgG1
shipped in   wet ice
storage temp.   2-8°C
Gene Information   human ... AGR3(155465)
biological source   mouse
conjugate   unconjugated
NCBI accession no.   NM_176813.3

Description

Immunogen

Purified human AGR3 protein

General description

The antibody AGR3.2 recognizes epitope QYSQALKKV within AGR3 (AG3) protein (19-20 kDa); secreted cytoplasmic protein which can serve as a marker of carcinogenesis.

Physical form

Solution in phosphate buffered saline, pH 7.4, with 15 mM sodium azide.

Price and Availability


Antibody Explorer

Explore with Confidence
Safety & Documentation

Safety Information

RIDADR  NONH for all modes of transport
Protocols & Articles

Related Content

Antibody Explorer | Buy Primary & Secondary Antibodies

Monoclonal and polyclonal primary antibodies are focused on cell biology, neurobiology and molecular biology. Secondary antibodies targeting multiple host’s IgG are conjugated to alkaline phosphatase...
Keywords: Amplification, Buffers, Cell biology, Enzyme-linked immunosorbent assay, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Molecular biology, Phosphorylations, Purification, Western blot

Related Products

Technical Service:

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Bulk Ordering & Pricing:

Need larger quantities for your development, manufacturing or research applications?